Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
Chromosome Location
Chr.5: 13690328 - 14011829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001369.2 NP_001360.1 68 - 69 11804 77
XM_005248262.3 XP_005248319.2 68 - 69 11887 77
XM_017009177.1 XP_016864666.1 68 - 69 11887 77
XM_017009178.1 XP_016864667.1 68 - 69 12690 77
XM_017009179.1 XP_016864668.1 62 - 63 10984 77
XM_017009180.1 XP_016864669.1 68 - 69 11887 77
XM_017009185.1 XP_016864674.1 40 - 41 7183 77
XM_017009186.1 XP_016864675.1 37 - 38 6599 77
XM_017009188.1 XP_016864677.1 34 - 35 7415 77
GenBank mRNA AB046823.2 - 16 - 17 2868 77
AB290164.1 - 21 - 22 3807 77
AK128600.1 - 10 - 11 1807 77
AY045575.1 - 68 - 69 11804 77

View Full Product Details