The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001369.2 | NP_001360.1 | 74 - 75 | 12948 | 65 |
XM_005248262.3 | XP_005248319.2 | 74 - 75 | 13031 | 65 | |
XM_017009178.1 | XP_016864667.1 | 74 - 75 | 13834 | 65 | |
XM_017009179.1 | XP_016864668.1 | 68 - 69 | 12128 | 65 | |
XM_017009185.1 | XP_016864674.1 | 46 - 47 | 8327 | 65 | |
XM_017009186.1 | XP_016864675.1 | 43 - 44 | 7743 | 65 | |
XM_017009188.1 | XP_016864677.1 | 40 - 41 | 8559 | 65 | |
GenBank mRNA | AB046823.2 | - | 22 - 23 | 4012 | 65 |
AB290164.1 | - | 27 - 28 | 4951 | 65 | |
AK026756.1 | - | 2 - 3 | 304 | 65 | |
AK128600.1 | - | 16 - 17 | 2951 | 65 | |
AY045575.1 | - | 74 - 75 | 12948 | 65 |