Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPAT4
Entrez Gene ID
137964
Gene Name
glycerol-3-phosphate acyltransferase 4
Gene Aliases
1-AGPAT 6, AGPAT6, LPAAT-zeta, LPAATZ, TSARG7
Chromosome Location
Chr.8: 41578188 - 41625001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178819.3 NP_848934.1 10 - 11 1999 90
XM_011544392.2 XP_011542694.1 11 - 12 2122 90
XM_011544393.1 XP_011542695.1 10 - 11 2011 90
XM_011544394.1 XP_011542696.1 10 - 11 2048 90
XM_017013042.1 XP_016868531.1 11 - 12 2149 90
XM_017013043.1 XP_016868532.1 11 - 12 2020 90
GenBank mRNA AF406612.1 - 10 - 11 1987 90
AL834255.1 - 6 - 7 484 90
AY358670.1 - 9 - 10 1136 90
AY513610.1 - 9 - 10 1219 90
BC051377.1 - 9 - 10 1179 90
BC061884.1 - 9 - 10 1179 90

View Full Product Details