The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206897.1 | NP_001193826.1 | 10 - 11 | 1453 | 85 |
NM_003888.3 | NP_003879.2 | 9 - 10 | 1356 | 85 | |
NM_170696.2 | NP_733797.1 | 8 - 9 | 1242 | 85 | |
NM_170697.2 | NP_733798.1 | 7 - 8 | 960 | 85 | |
GenBank mRNA | AB015226.1 | - | 9 - 10 | 1126 | 85 |
AB015227.1 | - | 7 - 8 | 889 | 85 | |
AB015228.1 | - | 7 - 8 | 851 | 85 | |
AK128709.1 | - | 9 - 10 | 1851 | 85 | |
AK294981.1 | - | 9 - 10 | 1056 | 85 | |
AK303057.1 | - | 10 - 11 | 1198 | 85 | |
AL110299.1 | - | 1 - 2 | 904 | 85 | |
BC030589.2 | - | 8 - 9 | 1000 | 85 |