Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM9B
Entrez Gene ID
171483
Gene Name
family with sequence similarity 9 member B
Gene Aliases
TEX39B
Chromosome Location
Chr.X: 9024232 - 9197176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205849.3 NP_995321.1 8 - 9 949 122
XM_011545464.2 XP_011543766.1 15 - 16 1522 122
GenBank mRNA AF494344.1 - 8 - 9 948 122
AK097501.1 - 7 - 8 893 122
AK128470.1 - 8 - 9 3310 122
BC120955.1 - 7 - 8 648 122
BC120956.1 - 7 - 8 648 122

View Full Product Details