The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308007.1 | NP_001294936.1 | 6 - 7 | 1483 | 106 |
NM_003797.4 | NP_003788.2 | 6 - 7 | 1483 | 106 | |
XM_005274373.2 | XP_005274430.1 | 6 - 7 | 1323 | 106 | |
XM_011545330.1 | XP_011543632.1 | 6 - 7 | 1323 | 106 | |
XM_011545331.1 | XP_011543633.1 | 6 - 7 | 1323 | 106 | |
XM_017018512.1 | XP_016874001.1 | 6 - 7 | 1323 | 106 | |
XM_017018513.1 | XP_016874002.1 | 6 - 7 | 1323 | 106 | |
GenBank mRNA | AF070418.1 | - | 6 - 7 | 865 | 106 |
AF078933.1 | - | 6 - 7 | 629 | 106 | |
AF080227.1 | - | 6 - 7 | 981 | 106 | |
AF099032.1 | - | 6 - 7 | 727 | 106 | |
AK292120.1 | - | 6 - 7 | 1092 | 106 | |
AK301265.1 | - | 6 - 7 | 599 | 106 | |
BC047672.2 | - | 6 - 7 | 596 | 106 | |
BC068995.1 | - | 6 - 7 | 644 | 106 | |
U90651.1 | - | 6 - 7 | 727 | 106 |