Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PQLC1
Entrez Gene ID
80148
Gene Name
PQ loop repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.18: 79902420 - 79951665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146343.1 NP_001139815.1 2 - 3 371 101
NM_001146345.1 NP_001139817.1 2 - 3 371 101
NM_025078.4 NP_079354.2 2 - 3 371 101
XM_005266770.2 XP_005266827.1 2 - 3 383 101
GenBank mRNA AK301881.1 - 2 - 3 371 101
AK303818.1 - 1 - 2 153 101
BC030140.2 - 2 - 3 335 101
DB475520.1 - 2 - 3 371 101

View Full Product Details