Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PQLC1
Entrez Gene ID
80148
Gene Name
PQ loop repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.18: 79902420 - 79951665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025078.4 NP_079354.2 3 - 4 513 75
XM_005266770.2 XP_005266827.1 3 - 4 525 75
XM_017025997.1 XP_016881486.1 4 - 5 600 75
XM_017025999.1 XP_016881488.1 4 - 5 600 75
XM_017026000.1 XP_016881489.1 4 - 5 600 75
XM_017026003.1 XP_016881492.1 4 - 5 600 75
GenBank mRNA AK123870.1 - 3 - 4 384 75
AK126188.1 - 1 - 2 1000 75
BC030140.2 - 3 - 4 477 75

View Full Product Details