Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- APBA2
- Entrez Gene ID
-
321
- Gene Name
- amyloid beta precursor protein binding family A member 2
- Gene Aliases
-
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L
- Chromosome Location
-
Chr.15: 28885479 - 29118315 on Build GRCh38
View Full Product Details