Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA3
Entrez Gene ID
3909
Gene Name
laminin subunit alpha 3
Gene Aliases
BM600, E170, LAMNA, LOCS
Chromosome Location
Chr.18: 23689443 - 23955066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000227.4 NP_000218.3 14 - 15 1814 64
NM_001127717.2 NP_001121189.2 50 - 51 6689 64
NM_001127718.2 NP_001121190.2 13 - 14 1646 64
NM_198129.2 NP_937762.2 51 - 52 6857 64
XM_011525978.2 XP_011524280.1 51 - 52 6884 64
XM_011525979.2 XP_011524281.1 51 - 52 6875 64
XM_011525980.2 XP_011524282.1 51 - 52 6866 64
XM_011525981.2 XP_011524283.1 50 - 51 6752 64
XM_011525982.2 XP_011524284.1 51 - 52 6884 64
XM_017025743.1 XP_016881232.1 34 - 35 4568 64
XM_017025744.1 XP_016881233.1 24 - 25 3249 64
GenBank mRNA AB107369.1 - 51 - 52 6616 64
AB208853.1 - 6 - 7 842 64
AY327114.1 - 13 - 14 1621 64
AY327115.1 - 51 - 52 6616 64
AY327116.1 - 50 - 51 6448 64
BC043618.1 - 8 - 9 1194 64
L34155.1 - 14 - 15 1756 64
X84900.1 - 16 - 17 2035 64
X85107.1 - 14 - 15 1807 64
X85108.1 - 16 - 17 2035 64

View Full Product Details