The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000227.4 | NP_000218.3 | 22 - 23 | 2972 | 130 |
NM_001127717.2 | NP_001121189.2 | 58 - 59 | 7847 | 130 | |
NM_001127718.2 | NP_001121190.2 | 21 - 22 | 2804 | 130 | |
NM_198129.2 | NP_937762.2 | 59 - 60 | 8015 | 130 | |
XM_011525978.2 | XP_011524280.1 | 59 - 60 | 8042 | 130 | |
XM_011525979.2 | XP_011524281.1 | 59 - 60 | 8033 | 130 | |
XM_011525980.2 | XP_011524282.1 | 59 - 60 | 8024 | 130 | |
XM_011525981.2 | XP_011524283.1 | 58 - 59 | 7910 | 130 | |
XM_011525982.2 | XP_011524284.1 | 57 - 58 | 7745 | 130 | |
XM_017025743.1 | XP_016881232.1 | 42 - 43 | 5726 | 130 | |
XM_017025744.1 | XP_016881233.1 | 32 - 33 | 4407 | 130 | |
GenBank mRNA | AB107369.1 | - | 59 - 60 | 7774 | 130 |
AB208853.1 | - | 14 - 15 | 2000 | 130 | |
AY327114.1 | - | 21 - 22 | 2779 | 130 | |
AY327115.1 | - | 59 - 60 | 7774 | 130 | |
AY327116.1 | - | 58 - 59 | 7606 | 130 | |
BC043618.1 | - | 16 - 17 | 2352 | 130 | |
L34155.1 | - | 22 - 23 | 2914 | 130 | |
X84900.1 | - | 24 - 25 | 3193 | 130 |