The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000227.4 | NP_000218.3 | 28 - 29 | 3779 | 140 |
NM_001127717.2 | NP_001121189.2 | 64 - 65 | 8654 | 140 | |
NM_001127718.2 | NP_001121190.2 | 27 - 28 | 3611 | 140 | |
NM_198129.2 | NP_937762.2 | 65 - 66 | 8822 | 140 | |
XM_011525978.2 | XP_011524280.1 | 65 - 66 | 8849 | 140 | |
XM_011525979.2 | XP_011524281.1 | 65 - 66 | 8840 | 140 | |
XM_011525980.2 | XP_011524282.1 | 65 - 66 | 8831 | 140 | |
XM_011525981.2 | XP_011524283.1 | 64 - 65 | 8717 | 140 | |
XM_011525982.2 | XP_011524284.1 | 63 - 64 | 8552 | 140 | |
XM_017025743.1 | XP_016881232.1 | 48 - 49 | 6533 | 140 | |
XM_017025744.1 | XP_016881233.1 | 38 - 39 | 5214 | 140 | |
GenBank mRNA | AB107369.1 | - | 65 - 66 | 8581 | 140 |
AB208853.1 | - | 20 - 21 | 2807 | 140 | |
AY327114.1 | - | 27 - 28 | 3586 | 140 | |
AY327115.1 | - | 65 - 66 | 8581 | 140 | |
AY327116.1 | - | 64 - 65 | 8413 | 140 | |
BC043618.1 | - | 22 - 23 | 3159 | 140 | |
L34155.1 | - | 28 - 29 | 3721 | 140 | |
X84900.1 | - | 30 - 31 | 4000 | 140 |