Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMA3
Entrez Gene ID
3909
Gene Name
laminin subunit alpha 3
Gene Aliases
BM600, E170, LAMNA, LOCS
Chromosome Location
Chr.18: 23689443 - 23955066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000227.4 NP_000218.3 34 - 35 4708 95
NM_001127717.2 NP_001121189.2 70 - 71 9583 95
NM_001127718.2 NP_001121190.2 33 - 34 4540 95
NM_198129.2 NP_937762.2 71 - 72 9751 95
XM_011525978.2 XP_011524280.1 71 - 72 9778 95
XM_011525979.2 XP_011524281.1 71 - 72 9769 95
XM_011525980.2 XP_011524282.1 71 - 72 9760 95
XM_011525981.2 XP_011524283.1 70 - 71 9646 95
XM_011525982.2 XP_011524284.1 69 - 70 9481 95
XM_017025743.1 XP_016881232.1 54 - 55 7462 95
XM_017025744.1 XP_016881233.1 44 - 45 6143 95
GenBank mRNA AB107369.1 - 71 - 72 9510 95
AB208853.1 - 26 - 27 3736 95
AY327114.1 - 33 - 34 4515 95
AY327115.1 - 71 - 72 9510 95
AY327116.1 - 70 - 71 9342 95
BC043618.1 - 28 - 29 4088 95
L34155.1 - 34 - 35 4650 95
X84900.1 - 36 - 37 4929 95

View Full Product Details