Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA3
Entrez Gene ID
3909
Gene Name
laminin subunit alpha 3
Gene Aliases
BM600, E170, LAMNA, LOCS
Chromosome Location
Chr.18: 23689443 - 23955066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000227.4 NP_000218.3 4 - 5 503 79
NM_001127717.2 NP_001121189.2 41 - 42 5546 79
NM_001127718.2 NP_001121190.2 4 - 5 503 79
NM_198129.2 NP_937762.2 41 - 42 5546 79
XM_011525978.2 XP_011524280.1 41 - 42 5573 79
XM_011525979.2 XP_011524281.1 41 - 42 5564 79
XM_011525980.2 XP_011524282.1 41 - 42 5555 79
XM_011525981.2 XP_011524283.1 40 - 41 5441 79
XM_011525982.2 XP_011524284.1 41 - 42 5573 79
XM_017025743.1 XP_016881232.1 24 - 25 3257 79
XM_017025744.1 XP_016881233.1 14 - 15 1938 79
GenBank mRNA AB107369.1 - 41 - 42 5305 79
AY327114.1 - 4 - 5 478 79
AY327115.1 - 41 - 42 5305 79
AY327116.1 - 41 - 42 5305 79
DA862923.1 - 4 - 5 503 79
L34155.1 - 4 - 5 445 79
X84900.1 - 6 - 7 724 79
X85107.1 - 4 - 5 496 79
X85108.1 - 6 - 7 724 79

View Full Product Details