The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014939.3 | NP_055754.2 | 28 - 29 | 4441 | 154 |
XM_005258233.2 | XP_005258290.1 | 28 - 29 | 4534 | 154 | |
XM_005258235.3 | XP_005258292.1 | 23 - 24 | 3315 | 154 | |
XM_011525884.2 | XP_011524186.1 | 27 - 28 | 4388 | 154 | |
XM_017025615.1 | XP_016881104.1 | 28 - 29 | 4033 | 154 | |
XM_017025617.1 | XP_016881106.1 | 27 - 28 | 4385 | 154 | |
GenBank mRNA | AB023229.1 | - | 28 - 29 | 4134 | 154 |
AK001662.1 | - | 20 - 21 | 2800 | 154 | |
AL136749.1 | - | 29 - 30 | 3966 | 154 | |
BC127109.1 | - | 28 - 29 | 3995 | 154 | |
BC127110.1 | - | 28 - 29 | 3995 | 154 |