The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161520.1 | NP_001154992.1 | 8 - 9 | 1453 | 130 |
NM_006348.3 | NP_006339.3 | 8 - 9 | 1453 | 130 | |
NM_181733.2 | NP_859422.2 | 8 - 9 | 1453 | 130 | |
GenBank mRNA | AF058718.1 | - | 8 - 9 | 979 | 130 |
AK314270.1 | - | 8 - 9 | 1451 | 130 | |
BC068540.1 | - | 8 - 9 | 1200 | 130 |