The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306151.1 | NP_001293080.1 | 7 - 8 | 768 | 121 |
NM_014395.2 | NP_055210.2 | 7 - 8 | 777 | 121 | |
XM_011531840.2 | XP_011530142.1 | 7 - 8 | 771 | 121 | |
XM_011531843.2 | XP_011530145.1 | 5 - 6 | 566 | 121 | |
GenBank mRNA | AF161551.1 | - | 7 - 8 | 752 | 121 |
AF163254.1 | - | 7 - 8 | 758 | 121 | |
AF178987.1 | - | 7 - 8 | 732 | 121 | |
AF186022.1 | - | 7 - 8 | 768 | 121 | |
BC012924.1 | - | 7 - 8 | 702 | 121 |