Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS2
Entrez Gene ID
57508
Gene Name
integrator complex subunit 2
Gene Aliases
INT2, KIAA1287
Chromosome Location
Chr.17: 61865367 - 61928029 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011525058.2 XP_011523360.1 1 - 2 371 87

View Full Product Details