The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020748.2 | NP_065799.1 | 23 - 24 | 3349 | 92 |
NR_026641.1 | 23 - 24 | 3630 | 92 | ||
XM_006721987.3 | XP_006722050.1 | 23 - 24 | 3356 | 92 | |
XM_011525058.2 | XP_011523360.1 | 23 - 24 | 3636 | 92 | |
GenBank mRNA | AB033113.1 | - | 23 - 24 | 3577 | 92 |
AK299032.1 | - | 23 - 24 | 3605 | 92 | |
AK308307.1 | - | 23 - 24 | 3314 | 92 | |
AL136800.1 | - | 23 - 24 | 3349 | 92 | |
CR533582.1 | - | 22 - 23 | 3273 | 92 |