The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006437.3 | NP_006428.2 | 31 - 32 | 4852 | 65 |
XM_011534931.1 | XP_011533233.1 | 30 - 31 | 4902 | 65 | |
XM_011534932.2 | XP_011533234.1 | 31 - 32 | 5069 | 65 | |
GenBank mRNA | AF057160.1 | - | 31 - 32 | 4798 | 65 |
AF158255.1 | - | 31 - 32 | 4852 | 65 | |
CA308288.1 | - | 3 - 4 | 65 | ||
D79999.2 | - | 30 - 31 | 4751 | 65 |