Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARP4
Entrez Gene ID
143
Gene Name
poly(ADP-ribose) polymerase family member 4
Gene Aliases
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193
Chromosome Location
Chr.13: 24420930 - 24512993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006437.3 NP_006428.2 9 - 10 1162 89
XM_011534931.1 XP_011533233.1 8 - 9 1212 89
XM_011534932.2 XP_011533234.1 9 - 10 1379 89
GenBank mRNA AF057160.1 - 9 - 10 1108 89
AF158255.1 - 9 - 10 1162 89
AK304399.1 - 9 - 10 1130 89
D79999.2 - 8 - 9 1061 89

View Full Product Details