The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171993.1 | NP_001165464.1 | 9 - 10 | 926 | 69 |
NM_002150.2 | NP_002141.1 | 7 - 8 | 450 | 69 | |
GenBank mRNA | AK057510.1 | - | 9 - 10 | 926 | 69 |
AK290826.1 | - | 7 - 8 | 446 | 69 | |
BC024287.1 | - | 7 - 8 | 432 | 69 | |
X72389.1 | - | 7 - 8 | 435 | 69 |