Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PIR
- Entrez Gene ID
-
8544
- Gene Name
- pirin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 15384799 - 15493588 on Build GRCh38
- Gene Symbol
- PIR-FIGF
- Entrez Gene ID
-
100532742
- Gene Name
- PIR-FIGF readthrough
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 15345591 - 15491309 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037859.2 |
|
2 - 3 |
246 |
87 |
GenBank mRNA |
AX740234.1 |
- |
2 - 3 |
954 |
87 |
View Full Product Details