Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAPP1
Entrez Gene ID
27071
Gene Name
dual adaptor of phosphotyrosine and 3-phosphoinositides 1
Gene Aliases
BAM32
Chromosome Location
Chr.4: 99816824 - 99872290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306151.1 NP_001293080.1 2 - 3 304 142
NM_014395.2 NP_055210.2 2 - 3 313 142
XM_011531840.2 XP_011530142.1 2 - 3 307 142
XM_011531842.1 XP_011530144.1 2 - 3 307 142
XM_017008023.1 XP_016863512.1 2 - 3 307 142
XM_017008024.1 XP_016863513.1 2 - 3 307 142
GenBank mRNA AF161551.1 - 2 - 3 288 142
AF163254.1 - 2 - 3 294 142
AF178987.1 - 2 - 3 268 142
AF186022.1 - 2 - 3 304 142
AK301355.1 - 2 - 3 287 142
BC012924.1 - 2 - 3 238 142
DN995752.1 - 2 - 3 324 142

View Full Product Details