The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143990.1 | NP_001137462.1 | 7 - 8 | 1104 | 57 |
NM_001143991.1 | NP_001137463.1 | 7 - 8 | 1114 | 57 | |
NM_001143992.1 | NP_001137464.1 | 7 - 8 | 1163 | 57 | |
NM_018081.2 | NP_060551.2 | 6 - 7 | 1249 | 57 | |
XM_011523952.2 | XP_011522254.1 | 6 - 7 | 890 | 57 | |
GenBank mRNA | AK001247.1 | - | 6 - 7 | 1121 | 57 |
AK056669.1 | - | 6 - 7 | 2918 | 57 | |
AK225273.1 | - | 5 - 6 | 1022 | 57 | |
AK225411.1 | - | 6 - 7 | 1121 | 57 | |
AY766322.1 | - | 7 - 8 | 1114 | 57 | |
BC002336.2 | - | 7 - 8 | 1024 | 57 | |
DQ431240.1 | - | 7 - 8 | 1163 | 57 | |
DQ431241.1 | - | 7 - 8 | 1104 | 57 |