Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRAP53
Entrez Gene ID
55135
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
Chromosome Location
Chr.17: 7686071 - 7703502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143990.1 NP_001137462.1 9 - 10 1420 59
NM_001143991.1 NP_001137463.1 9 - 10 1430 59
NM_001143992.1 NP_001137464.1 9 - 10 1479 59
NM_018081.2 NP_060551.2 8 - 9 1565 59
XM_011523952.2 XP_011522254.1 8 - 9 1206 59
GenBank mRNA AK001247.1 - 8 - 9 1437 59
AK056669.1 - 8 - 9 3234 59
AK092370.1 - 9 - 10 2260 59
AK225273.1 - 7 - 8 1338 59
AK225411.1 - 8 - 9 1437 59
AY766322.1 - 9 - 10 1430 59
BC002336.2 - 9 - 10 1340 59
DQ431240.1 - 9 - 10 1479 59
DQ431241.1 - 9 - 10 1420 59

View Full Product Details