The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016426.6 | NP_057510.4 | 7 - 8 | 1638 | 139 |
XM_005261627.3 | XP_005261684.1 | 7 - 8 | 2498 | 139 | |
XM_011530211.2 | XP_011528513.1 | 6 - 7 | 2440 | 139 | |
XM_017028815.1 | XP_016884304.1 | 6 - 7 | 1888 | 139 | |
XM_017028816.1 | XP_016884305.1 | 7 - 8 | 2500 | 139 | |
GenBank mRNA | AF223408.1 | - | 6 - 7 | 1439 | 139 |
AK303087.1 | - | 6 - 7 | 1409 | 139 | |
AK315550.1 | - | 7 - 8 | 1464 | 139 | |
AY927426.1 | - | 1 - 2 | 378 | 139 | |
AY927427.1 | - | 1 - 2 | 378 | 139 | |
BC006325.2 | - | 6 - 7 | 1416 | 139 | |
BC075828.1 | - | 7 - 8 | 1539 | 139 | |
CT841519.1 | - | 6 - 7 | 1382 | 139 |