The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271213.1 | NP_001258142.1 | 10 - 11 | 1489 | 78 |
NM_021199.3 | NP_067022.1 | 9 - 10 | 1682 | 78 | |
GenBank mRNA | AF042284.1 | - | 9 - 10 | 1372 | 78 |
AF118085.1 | - | 8 - 9 | 1203 | 78 | |
AF151802.1 | - | 9 - 10 | 1366 | 78 | |
AI240325.1 | - | 1 - 2 | 78 | ||
AK130140.1 | - | 10 - 11 | 1489 | 78 | |
AK222463.1 | - | 9 - 10 | 1381 | 78 | |
AK313111.1 | - | 9 - 10 | 1361 | 78 | |
BC016836.1 | - | 9 - 10 | 1337 | 78 |