Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRD1
Entrez Gene ID
3824
Gene Name
killer cell lectin like receptor D1
Gene Aliases
CD94
Chromosome Location
Chr.12: 10238385 - 10329607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114396.1 NP_001107868.1 2 - 3 205 75
NM_002262.3 NP_002253.1 1 - 2 268 75
XM_006719067.3 XP_006719130.1 1 - 2 257 75
XM_011520650.2 XP_011518952.1 2 - 3 199 75
XM_011520651.2 XP_011518953.1 1 - 2 256 75
XM_017019285.1 XP_016874774.1 1 - 2 260 75
XM_017019286.1 XP_016874775.1 2 - 3 190 75
XM_017019287.1 XP_016874776.1 2 - 3 210 75
XM_017019290.1 XP_016874779.1 3 - 4 2066 75
XM_017019291.1 XP_016874780.1 2 - 3 192 75
GenBank mRNA AF498040.1 - 1 - 2 136 75
AF498041.1 - 1 - 2 136 75
AK291506.1 - 2 - 3 210 75
AY227806.1 - 2 - 3 205 75
AY327501.1 - 1 - 2 136 75
BC028009.1 - 1 - 2 81 75
U30610.1 - 1 - 2 92 75

View Full Product Details