Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPR2
Entrez Gene ID
3709
Gene Name
inositol 1,4,5-trisphosphate receptor type 2
Gene Aliases
ANHD, CFAP48, INSP3R2, IP3R2
Chromosome Location
Chr.12: 26335337 - 26833198 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002223.3 NP_002214.2 31 - 32 4675 109
XM_017019266.1 XP_016874755.1 32 - 33 4734 109
XM_017019267.1 XP_016874756.1 32 - 33 4258 109
XM_017019268.1 XP_016874757.1 31 - 32 4135 109
XM_017019269.1 XP_016874758.1 32 - 33 4734 109
GenBank mRNA D26350.1 - 31 - 32 4676 109

View Full Product Details