Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPR2
Entrez Gene ID
3709
Gene Name
inositol 1,4,5-trisphosphate receptor type 2
Gene Aliases
ANHD, CFAP48, INSP3R2, IP3R2
Chromosome Location
Chr.12: 26335337 - 26833198 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002223.3 NP_002214.2 49 - 50 7354 127
XM_017019266.1 XP_016874755.1 50 - 51 7413 127
XM_017019267.1 XP_016874756.1 50 - 51 6937 127
XM_017019268.1 XP_016874757.1 49 - 50 6814 127
GenBank mRNA AK307686.1 - 12 - 13 1765 127
D26350.1 - 49 - 50 7355 127

View Full Product Details