The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002223.3 | NP_002214.2 | 5 - 6 | 934 | 107 |
XM_017019266.1 | XP_016874755.1 | 5 - 6 | 933 | 107 | |
XM_017019267.1 | XP_016874756.1 | 5 - 6 | 457 | 107 | |
XM_017019269.1 | XP_016874758.1 | 5 - 6 | 933 | 107 | |
GenBank mRNA | AK307712.1 | - | 5 - 6 | 810 | 107 |
AK310504.1 | - | 5 - 6 | 674 | 107 | |
AY429550.1 | - | 4 - 5 | 451 | 107 | |
BX495410.1 | - | 1 - 2 | 154 | 107 | |
D26350.1 | - | 5 - 6 | 935 | 107 |