Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP5S
Entrez Gene ID
27109
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B)
Gene Aliases
ATPW, FB, HSU79253
Chromosome Location
Chr.14: 50311752 - 50326715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003803.2 NP_001003803.1 4 - 5 1222 118
NR_033761.1 5 - 6 1258 118
XM_005267537.3 XP_005267594.1 4 - 5 1129 118
XM_011536649.2 XP_011534951.1 4 - 5 1111 118
XM_011536652.1 XP_011534954.1 4 - 5 511 118
XM_011536653.2 XP_011534955.1 5 - 6 553 118
XM_017021220.1 XP_016876709.1 4 - 5 593 118
GenBank mRNA AK290008.1 - 4 - 5 910 118
AY052377.1 - 4 - 5 645 118
BU189283.1 - 4 - 5 533 118

View Full Product Details