The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167856.1 | NP_001161328.1 | 11 - 12 | 1534 | 126 |
NM_018183.3 | NP_060653.3 | 11 - 12 | 1531 | 126 | |
XM_005253572.3 | XP_005253629.1 | 12 - 13 | 1672 | 126 | |
XM_005253573.4 | XP_005253630.1 | 13 - 14 | 2082 | 126 | |
XM_005253575.3 | XP_005253632.1 | 12 - 13 | 1669 | 126 | |
XM_005253576.3 | XP_005253633.1 | 13 - 14 | 1713 | 126 | |
XM_006719473.3 | XP_006719536.1 | 14 - 15 | 1838 | 126 | |
XM_006719474.3 | XP_006719537.1 | 13 - 14 | 1791 | 126 | |
XM_011538533.2 | XP_011536835.1 | 14 - 15 | 1832 | 126 | |
XM_011538534.2 | XP_011536836.1 | 11 - 12 | 1570 | 126 | |
XM_011538535.2 | XP_011536837.1 | 11 - 12 | 1572 | 126 | |
XM_017019556.1 | XP_016875045.1 | 12 - 13 | 1656 | 126 | |
XM_017019557.1 | XP_016875046.1 | 12 - 13 | 1663 | 126 | |
XM_017019558.1 | XP_016875047.1 | 13 - 14 | 1726 | 126 | |
XM_017019559.1 | XP_016875048.1 | 12 - 13 | 1793 | 126 | |
GenBank mRNA | AB014772.1 | - | 11 - 12 | 1531 | 126 |
AK001695.1 | - | 12 - 13 | 1669 | 126 | |
AK122703.1 | - | 13 - 14 | 2087 | 126 | |
AK307488.1 | - | 11 - 12 | 1570 | 126 | |
AY364255.1 | - | 11 - 12 | 1533 | 126 | |
BC030544.1 | - | 12 - 13 | 1665 | 126 | |
BC133704.1 | - | 11 - 12 | 1534 | 126 |