The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167856.1 | NP_001161328.1 | 29 - 30 | 3847 | 98 |
NM_018183.3 | NP_060653.3 | 29 - 30 | 3844 | 98 | |
XM_005253572.3 | XP_005253629.1 | 30 - 31 | 3985 | 98 | |
XM_005253573.4 | XP_005253630.1 | 31 - 32 | 4395 | 98 | |
XM_005253575.3 | XP_005253632.1 | 30 - 31 | 3982 | 98 | |
XM_005253576.3 | XP_005253633.1 | 31 - 32 | 4026 | 98 | |
XM_006719473.3 | XP_006719536.1 | 32 - 33 | 4151 | 98 | |
XM_006719474.3 | XP_006719537.1 | 31 - 32 | 4104 | 98 | |
XM_011538533.2 | XP_011536835.1 | 32 - 33 | 4145 | 98 | |
XM_011538534.2 | XP_011536836.1 | 29 - 30 | 3883 | 98 | |
XM_011538535.2 | XP_011536837.1 | 29 - 30 | 3885 | 98 | |
XM_017019556.1 | XP_016875045.1 | 30 - 31 | 3969 | 98 | |
XM_017019557.1 | XP_016875046.1 | 30 - 31 | 3976 | 98 | |
XM_017019558.1 | XP_016875047.1 | 31 - 32 | 4039 | 98 | |
XM_017019559.1 | XP_016875048.1 | 30 - 31 | 4106 | 98 | |
GenBank mRNA | AB014772.1 | - | 29 - 30 | 3844 | 98 |
AK001563.1 | - | 6 - 7 | 590 | 98 | |
AK074256.1 | - | 13 - 14 | 1414 | 98 | |
AK096864.1 | - | 12 - 13 | 1217 | 98 | |
BC133704.1 | - | 29 - 30 | 3847 | 98 |