Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBNO1
Entrez Gene ID
55206
Gene Name
strawberry notch homolog 1
Gene Aliases
MOP3, Sno
Chromosome Location
Chr.12: 123289109 - 123365204 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167856.1 NP_001161328.1 1 - 2 134 65
NM_018183.3 NP_060653.3 1 - 2 134 65
XM_005253572.3 XP_005253629.1 2 - 3 272 65
XM_005253573.4 XP_005253630.1 3 - 4 682 65
XM_005253575.3 XP_005253632.1 2 - 3 272 65
XM_005253576.3 XP_005253633.1 3 - 4 316 65
XM_006719473.3 XP_006719536.1 4 - 5 438 65
XM_006719474.3 XP_006719537.1 3 - 4 391 65
XM_011538533.2 XP_011536835.1 4 - 5 435 65
XM_017019556.1 XP_016875045.1 2 - 3 262 65
XM_017019557.1 XP_016875046.1 2 - 3 272 65
XM_017019558.1 XP_016875047.1 3 - 4 335 65
GenBank mRNA AB014772.1 - 1 - 2 134 65
AK001695.1 - 2 - 3 272 65
AK122703.1 - 3 - 4 687 65
AY364255.1 - 1 - 2 136 65
BC030544.1 - 2 - 3 271 65
BC133704.1 - 1 - 2 134 65

View Full Product Details