The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178078.1 | NP_001171549.1 | 1 - 2 | 95 | |
NM_003153.4 | NP_003144.3 | 1 - 2 | 292 | 76 | |
NR_033659.1 | 1 - 2 | 292 | 76 | ||
XM_011538703.2 | XP_011537005.1 | 1 - 2 | 312 | 76 | |
XM_011538707.2 | XP_011537009.1 | 1 - 2 | 95 | ||
XM_011538708.2 | XP_011537010.1 | 1 - 2 | 255 | 76 | |
GenBank mRNA | AK290431.1 | - | 1 - 2 | 95 | |
AK299094.1 | - | 1 - 2 | 228 | 76 | |
AK312471.1 | - | 1 - 2 | 158 | 76 | |
AK316236.1 | - | 1 - 2 | 69 | 76 | |
BC075852.1 | - | 1 - 2 | 93 | 76 | |
U16031.1 | - | 1 - 2 | 151 | 76 |