Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATL3
Entrez Gene ID
25923
Gene Name
atlastin GTPase 3
Gene Aliases
HSN1F
Chromosome Location
Chr.11: 63624082 - 63671974 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290048.1 NP_001276977.1 3 - 4 696 124
NM_015459.4 NP_056274.3 3 - 4 798 124
XM_006718493.1 XP_006718556.1 3 - 4 680 124
GenBank mRNA AK023383.1 - 3 - 4 403 124
AK097588.1 - 5 - 6 738 124
AK301910.1 - 3 - 4 634 124
BC077727.1 - 3 - 4 689 124
CR936784.1 - 3 - 4 681 124

View Full Product Details