The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002211.3 | NP_002202.2 | 3 - 4 | 368 | 81 |
NM_033668.2 | NP_391988.1 | 2 - 3 | 147 | 81 | |
NM_133376.2 | NP_596867.1 | 3 - 4 | 283 | 81 | |
GenBank mRNA | AK291697.1 | - | 3 - 4 | 220 | 81 |
AL710802.1 | - | 2 - 3 | 159 | 81 | |
BC020057.1 | - | 3 - 4 | 212 | 81 | |
BX537407.1 | - | 3 - 4 | 222 | 81 | |
DB292597.1 | - | 3 - 4 | 223 | 81 |