The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005406.2 | NP_005397.1 | 23 - 24 | 3756 | 105 |
XM_011526137.2 | XP_011524439.1 | 12 - 13 | 1990 | 105 | |
GenBank mRNA | AB208965.1 | - | 15 - 16 | 1849 | 105 |
AK307388.1 | - | 23 - 24 | 3571 | 105 | |
BC113114.1 | - | 23 - 24 | 2831 | 105 | |
U43195.1 | - | 23 - 24 | 2815 | 105 |