Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATRNL1
Entrez Gene ID
26033
Gene Name
attractin like 1
Gene Aliases
ALP, bA338L11.1, bA454H24.1
Chromosome Location
Chr.10: 115093365 - 115948999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207303.4 NP_997186.1 21 - 22 3709 107
XM_011539587.1 XP_011537889.1 20 - 21 3566 107
XM_011539588.2 XP_011537890.1 21 - 22 3254 107
XM_011539589.2 XP_011537891.1 21 - 22 3704 107
XM_011539590.2 XP_011537892.1 21 - 22 5606 107
XM_011539591.2 XP_011537893.1 21 - 22 3577 107
XM_017016035.1 XP_016871524.1 21 - 22 3704 107
XM_017016037.1 XP_016871526.1 19 - 20 2708 107
XM_017016038.1 XP_016871527.1 19 - 20 10522 107
XM_017016039.1 XP_016871528.1 13 - 14 1873 107
XM_017016040.1 XP_016871529.1 10 - 11 1389 107
GenBank mRNA AB011106.2 - 15 - 16 2270 107
AK127277.1 - 21 - 22 3187 107
AK294917.1 - 5 - 6 573 107
AY442317.1 - 21 - 22 3323 107

View Full Product Details