The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207303.4 | NP_997186.1 | 28 - 29 | 4401 | 135 |
XM_011539587.1 | XP_011537889.1 | 27 - 28 | 4258 | 135 | |
XM_011539588.2 | XP_011537890.1 | 28 - 29 | 3946 | 135 | |
XM_011539590.2 | XP_011537892.1 | 28 - 29 | 6298 | 135 | |
XM_011539591.2 | XP_011537893.1 | 28 - 29 | 4269 | 135 | |
XM_017016037.1 | XP_016871526.1 | 26 - 27 | 3400 | 135 | |
XM_017016038.1 | XP_016871527.1 | 26 - 27 | 11214 | 135 | |
XM_017016039.1 | XP_016871528.1 | 20 - 21 | 2565 | 135 | |
XM_017016040.1 | XP_016871529.1 | 17 - 18 | 2081 | 135 | |
GenBank mRNA | AB011106.2 | - | 22 - 23 | 2962 | 135 |
AK127277.1 | - | 28 - 29 | 3879 | 135 | |
AK294917.1 | - | 12 - 13 | 1265 | 135 | |
AK299592.1 | - | 8 - 9 | 798 | 135 | |
AY442317.1 | - | 28 - 29 | 4015 | 135 |