Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFXN2
Entrez Gene ID
118980
Gene Name
sideroflexin 2
Gene Aliases
-
Chromosome Location
Chr.10: 102714518 - 102739189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK055711.1 - 6 - 7 645 78

View Full Product Details