Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX25
Entrez Gene ID
83891
Gene Name
sorting nexin 25
Gene Aliases
MSTP043, SBBI31
Chromosome Location
Chr.4: 185204237 - 185363971 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317781.1 NP_001304710.1 8 - 9 1278 139
XM_005263267.4 XP_005263324.1 8 - 9 1721 139
XM_011532302.2 XP_011530604.1 10 - 11 1856 139
XM_011532303.2 XP_011530605.1 10 - 11 1853 139
XM_011532304.2 XP_011530606.1 9 - 10 1820 139
XM_011532305.2 XP_011530607.1 9 - 10 1757 139
XM_011532306.2 XP_011530608.1 10 - 11 1856 139
XM_011532307.2 XP_011530609.1 10 - 11 1856 139
XM_011532308.2 XP_011530610.1 9 - 10 1820 139
XM_017008680.1 XP_016864169.1 9 - 10 1820 139
XM_017008681.1 XP_016864170.1 10 - 11 1856 139
XM_017008682.1 XP_016864171.1 8 - 9 1721 139
XM_017008683.1 XP_016864172.1 7 - 8 765 139
XM_017008684.1 XP_016864173.1 9 - 10 1037 139
XM_017008685.1 XP_016864174.1 3 - 4 483 139
GenBank mRNA AF113223.1 - 2 - 3 163 139
AK095661.1 - 5 - 6 682 139
AY044656.1 - 1 - 2 78 139
AY601647.1 - 8 - 9 1278 139
BC029868.1 - 3 - 4 518 139

View Full Product Details