The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139076.2 | NP_620775.2 | 4 - 5 | 349 | 90 |
XM_005263280.4 | XP_005263337.1 | 5 - 6 | 482 | 90 | |
GenBank mRNA | AK022704.1 | - | 4 - 5 | 335 | 90 |
AY358576.1 | - | 4 - 5 | 300 | 90 | |
BC039573.1 | - | 4 - 5 | 296 | 90 | |
DA765424.1 | - | 4 - 5 | 349 | 90 | |
EF531340.1 | - | 4 - 5 | 284 | 90 |