The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297755.1 | NP_001284684.1 | 10 - 11 | 2280 | 143 |
NM_001297756.1 | NP_001284685.1 | 11 - 12 | 2494 | 143 | |
NM_001297757.1 | NP_001284686.1 | 10 - 11 | 2383 | 143 | |
NM_133636.3 | NP_598375.2 | 11 - 12 | 2481 | 143 | |
XM_005262711.1 | XP_005262768.1 | 11 - 12 | 2342 | 143 | |
XM_005262713.2 | XP_005262770.1 | 11 - 12 | 2571 | 143 | |
XM_006714076.2 | XP_006714139.1 | 11 - 12 | 2443 | 143 | |
XM_017007679.1 | XP_016863168.1 | 10 - 11 | 2141 | 143 | |
XM_017007680.1 | XP_016863169.1 | 10 - 11 | 2370 | 143 | |
XM_017007682.1 | XP_016863171.1 | 11 - 12 | 2562 | 143 | |
XM_017007683.1 | XP_016863172.1 | 9 - 10 | 1681 | 143 | |
GenBank mRNA | AF436845.1 | - | 11 - 12 | 2442 | 143 |
AK308395.1 | - | 10 - 11 | 2221 | 143 | |
AK312725.1 | - | 11 - 12 | 2458 | 143 | |
AL512702.1 | - | 9 - 10 | 1145 | 143 | |
BC111502.1 | - | 10 - 11 | 2303 | 143 |