Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HELQ
Entrez Gene ID
113510
Gene Name
helicase, POLQ-like
Gene Aliases
HEL308
Chromosome Location
Chr.4: 83407343 - 83455883 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297755.1 NP_001284684.1 5 - 6 1551 125
NM_133636.3 NP_598375.2 6 - 7 1752 125
XM_005262711.1 XP_005262768.1 6 - 7 1613 125
XM_005262713.2 XP_005262770.1 6 - 7 1842 125
XM_006714076.2 XP_006714139.1 6 - 7 1714 125
XM_011531580.2 XP_011529882.1 6 - 7 1714 125
XM_017007679.1 XP_016863168.1 5 - 6 1412 125
XM_017007680.1 XP_016863169.1 5 - 6 1641 125
XM_017007681.1 XP_016863170.1 6 - 7 1714 125
GenBank mRNA AF436845.1 - 6 - 7 1713 125
AK308395.1 - 5 - 6 1491 125
CK000270.1 - 3 - 4 352 125

View Full Product Details