Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SENP6
Entrez Gene ID
26054
Gene Name
SUMO1/sentrin specific peptidase 6
Gene Aliases
SSP1, SUSP1
Chromosome Location
Chr.6: 75601509 - 75718285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100409.2 NP_001093879.1 14 - 15 2944 70
NM_001304792.1 NP_001291721.1 14 - 15 2944 70
NM_015571.3 NP_056386.2 15 - 16 2965 70
XM_006715457.3 XP_006715520.1 16 - 17 2719 70
XM_011535746.2 XP_011534048.1 14 - 15 2517 70
XM_011535747.2 XP_011534049.1 14 - 15 2868 70
XM_017010719.1 XP_016866208.1 15 - 16 2607 70
XM_017010720.1 XP_016866209.1 15 - 16 1923 70
XM_017010721.1 XP_016866210.1 15 - 16 1931 70
XM_017010722.1 XP_016866211.1 16 - 17 2738 70
XM_017010723.1 XP_016866212.1 16 - 17 2715 70
XM_017010724.1 XP_016866213.1 16 - 17 3090 70
XM_017010725.1 XP_016866214.1 13 - 14 2850 70
GenBank mRNA AB018340.2 - 15 - 16 1993 70
AF196304.1 - 15 - 16 1949 70
AF306508.1 - 15 - 16 1964 70
AF307849.1 - 15 - 16 2024 70
AK001406.1 - 12 - 13 1619 70
AK021417.1 - 6 - 7 796 70
AK096455.1 - 14 - 15 2526 70
AK126921.1 - 14 - 15 2944 70
BC028583.1 - 14 - 15 2151 70

View Full Product Details