The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100409.2 | NP_001093879.1 | 19 - 20 | 3815 | 73 |
NM_015571.3 | NP_056386.2 | 20 - 21 | 3836 | 73 | |
XM_006715457.3 | XP_006715520.1 | 21 - 22 | 3590 | 73 | |
XM_011535746.2 | XP_011534048.1 | 19 - 20 | 3388 | 73 | |
XM_011535747.2 | XP_011534049.1 | 19 - 20 | 3739 | 73 | |
XM_017010719.1 | XP_016866208.1 | 20 - 21 | 3478 | 73 | |
XM_017010720.1 | XP_016866209.1 | 20 - 21 | 2794 | 73 | |
XM_017010721.1 | XP_016866210.1 | 20 - 21 | 2802 | 73 | |
XM_017010722.1 | XP_016866211.1 | 21 - 22 | 3609 | 73 | |
XM_017010723.1 | XP_016866212.1 | 21 - 22 | 3586 | 73 | |
XM_017010724.1 | XP_016866213.1 | 21 - 22 | 3961 | 73 | |
XM_017010725.1 | XP_016866214.1 | 18 - 19 | 3721 | 73 | |
GenBank mRNA | AB018340.2 | - | 20 - 21 | 2864 | 73 |
AF196304.1 | - | 20 - 21 | 2820 | 73 | |
AF306508.1 | - | 20 - 21 | 2835 | 73 | |
AF307849.1 | - | 20 - 21 | 2895 | 73 | |
AK021417.1 | - | 10 - 11 | 1239 | 73 | |
AK021949.1 | - | 4 - 5 | 759 | 73 | |
AK126921.1 | - | 19 - 20 | 3815 | 73 | |
BC028583.1 | - | 19 - 20 | 3022 | 73 |