The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100409.2 | NP_001093879.1 | 6 - 7 | 1540 | 111 |
NM_001304792.1 | NP_001291721.1 | 6 - 7 | 1540 | 111 | |
NM_015571.3 | NP_056386.2 | 7 - 8 | 1561 | 111 | |
XM_006715457.3 | XP_006715520.1 | 8 - 9 | 1315 | 111 | |
XM_011535746.2 | XP_011534048.1 | 6 - 7 | 1113 | 111 | |
XM_011535747.2 | XP_011534049.1 | 6 - 7 | 1464 | 111 | |
XM_017010719.1 | XP_016866208.1 | 7 - 8 | 1203 | 111 | |
XM_017010720.1 | XP_016866209.1 | 7 - 8 | 519 | 111 | |
XM_017010721.1 | XP_016866210.1 | 7 - 8 | 527 | 111 | |
XM_017010722.1 | XP_016866211.1 | 8 - 9 | 1334 | 111 | |
XM_017010723.1 | XP_016866212.1 | 8 - 9 | 1311 | 111 | |
XM_017010724.1 | XP_016866213.1 | 8 - 9 | 1686 | 111 | |
XM_017010725.1 | XP_016866214.1 | 5 - 6 | 1446 | 111 | |
GenBank mRNA | AB018340.2 | - | 7 - 8 | 589 | 111 |
AF196304.1 | - | 7 - 8 | 545 | 111 | |
AF306508.1 | - | 7 - 8 | 560 | 111 | |
AF307849.1 | - | 7 - 8 | 620 | 111 | |
AK001406.1 | - | 4 - 5 | 215 | 111 | |
AK096455.1 | - | 6 - 7 | 1122 | 111 | |
AK126921.1 | - | 6 - 7 | 1540 | 111 | |
BC028583.1 | - | 6 - 7 | 747 | 111 |